siRNA Details
| Gene Symbol: Car10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028296.3 | NP_082572.1 | 3 | 1194 | ||
| GenBank mRNA | AB080741.1 | - | 4 | 1150 | ||
| AK012302.1 | - | 4 | 1157 | |||
| AK035758.1 | - | 3 | 429 | |||
| AK038695.1 | - | 4 | 1074 | |||
| AK132222.1 | - | 3 | 1035 | |||
| AK162576.1 | - | 3 | 312 | |||
| BC017606.1 | - | 3 | 386 | 4507552 | ||
| BC066111.1 | - | 4 | 1150 | 30360743 | ||

Car10
Genomic Map