siRNA Details
| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004855.1 | - | 18 | 3014 | ||
| AK045033.1 | - | 19 | 3442 | |||
| AK050152.1 | - | 18 | 3025 | |||
| AK173193.1 | - | 7 | 7073 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stra6l| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004855.1 | - | 18 | 3014 | ||
| AK045033.1 | - | 19 | 3442 | |||
| AK050152.1 | - | 18 | 3025 | |||
| AK173193.1 | - | 7 | 7073 | |||