siRNA Details
| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004855.1 | - | 18 | 2138 | ||
| AK045033.1 | - | 19 | 2566 | |||
| AK050152.1 | - | 18 | 2149 | |||
| AK050348.1 | - | 18 | 2145 | |||
| AK173193.1 | - | 7 | 6197 | |||
| BC111889.1 | - | 18 | 2075 | 40060643 | ||
| BC112408.1 | - | 18 | 2075 | 40060646 | ||

Stra6l
Genomic Map