siRNA Details
| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033606.2 | NP_001028778.2 | 11, 12 | 1863 | ||
| NM_001136222.1 | NP_001129694.1 | 10, 11 | 1445 | |||
| NM_028817.3 | NP_083093.2 | 12, 13 | 2003 | |||
| GenBank mRNA | AK012088.1 | - | 11, 12 | 1806 | ||
| AK160839.1 | - | 10, 11 | 1389 | |||
| AK161708.1 | - | 12, 13 | 2004 | |||
| AK166723.1 | - | 12, 13 | 1941 | |||
| BC031529.1 | - | 10, 11 | 1588 | 4009004 | ||

Acsl3
Genomic Map