siRNA Details
| Gene Symbol: Lrrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028838.2 | NP_083114.2 | 9 | 1374 | ||
| GenBank mRNA | AJ428068.1 | - | 9 | 1373 | ||
| AK010252.1 | - | 9 | 1373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc2| Gene Symbol: Lrrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028838.2 | NP_083114.2 | 9 | 1374 | ||
| GenBank mRNA | AJ428068.1 | - | 9 | 1373 | ||
| AK010252.1 | - | 9 | 1373 | |||