siRNA Details
| Gene Symbol: Spatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028852.1 | NP_083128.1 | 2 | 892 | ||
| GenBank mRNA | AB032199.1 | - | 2 | 892 | ||
| AK006994.1 | - | 2 | 893 | |||
| BC116725.1 | - | 2 | 859 | |||
| BC118509.1 | - | 2 | 852 | 40106461 | ||

Spatc1
Genomic Map