siRNA Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028881.2 | NP_083157.1 | 10 | 1059 | ||
| GenBank mRNA | AK014553.1 | - | 5 | 1430 | ||
| AK079382.1 | - | 8 | 1807 | |||
| AK156192.1 | - | 10 | 1038 | |||
| AK169944.1 | - | 6 | 708 | |||
| BC023091.1 | - | 6 | 649 | 4023064 | ||
| BC023831.1 | - | 10 | 1054 | 5345301 | ||
| BC032183.1 | - | 10 | 1064 | 5310152 | ||
| BC033300.1 | - | 10 | 1054 | 4166411 | ||
| CV783724.1 | - | 7 | 563 | |||

Crtc2
Genomic Map