siRNA Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028881.2 | NP_083157.1 | 13 | 1970 | ||
| GenBank mRNA | AA276938.1 | - | 2 | 195 | ||
| AK014553.1 | - | 7 | 2077 | |||
| AK079382.1 | - | 10 | 2311 | |||
| AK156192.1 | - | 13 | 1949 | |||
| AK169944.1 | - | 9 | 1619 | |||
| BC019713.1 | - | 3 | 352 | |||
| BC023091.1 | - | 9 | 1560 | 4023064 | ||
| BC023831.1 | - | 13 | 1965 | 5345301 | ||
| BC032183.1 | - | 13 | 1975 | 5310152 | ||
| BC033300.1 | - | 13 | 1965 | 4166411 | ||

Crtc2
Genomic Map