siRNA Details
| Gene Symbol: Gle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028923.3 | NP_083199.1 | 8 | 1245 | ||
| GenBank mRNA | AK016671.1 | - | 8 | 1227 | ||
| AK028183.1 | - | 8 | 1229 | |||
| AK133888.1 | - | 8 | 1197 | |||
| AK145748.1 | - | 8 | 1221 | |||
| AK161015.1 | - | 8 | 1191 | |||
| AK161659.1 | - | 8 | 1228 | |||
| BC026797.1 | - | 8 | 1225 | 3990668 | ||

Gle1
Genomic Map