siRNA Details
| Gene Symbol: Lrrc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028973.2 | NP_083249.1 | 2 | 4105 | ||
| GenBank mRNA | AK162723.1 | - | 1 | 713 | ||
| BC050245.1 | - | 2 | 4077 | 6312567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc15| Gene Symbol: Lrrc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028973.2 | NP_083249.1 | 2 | 4105 | ||
| GenBank mRNA | AK162723.1 | - | 1 | 713 | ||
| BC050245.1 | - | 2 | 4077 | 6312567 | ||