siRNA Details
| Gene Symbol: 4930432K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029045.2 | NP_083321.2 | 5 | 696 | ||
| GenBank mRNA | AK208111.1 | - | 3 | 154 | ||
| BC075658.1 | - | 5 | 575 | 30607069 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930432K21Rik| Gene Symbol: 4930432K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029045.2 | NP_083321.2 | 5 | 696 | ||
| GenBank mRNA | AK208111.1 | - | 3 | 154 | ||
| BC075658.1 | - | 5 | 575 | 30607069 | ||