siRNA Details
| Gene Symbol: 4930432K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163752.1 | NP_001157224.1 | 8 | 2159 | ||
| NM_029045.2 | NP_083321.2 | 9 | 2264 | |||
| GenBank mRNA | AK015293.1 | - | 8 | 1946 | ||
| BC075658.1 | - | 9 | 2143 | 30607069 | ||

4930432K21Rik
Genomic Map