siRNA Details
| Gene Symbol: Rnase10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162863.1 | NP_001156335.1 | 2 | 859 | ||
| NM_029145.3 | NP_083421.2 | 2 | 968 | |||
| GenBank mRNA | AK015573.1 | - | 2 | 860 | ||
| AY226990.1 | - | 2 | 968 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase10| Gene Symbol: Rnase10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162863.1 | NP_001156335.1 | 2 | 859 | ||
| NM_029145.3 | NP_083421.2 | 2 | 968 | |||
| GenBank mRNA | AK015573.1 | - | 2 | 860 | ||
| AY226990.1 | - | 2 | 968 | |||