siRNA Details
| Gene Symbol: Rnase10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162863.1 | NP_001156335.1 | 2 | 380 | ||
| NM_029145.3 | NP_083421.2 | 2 | 489 | |||
| GenBank mRNA | AK015573.1 | - | 2 | 381 | ||
| AY226990.1 | - | 2 | 489 | |||
| BC141116.1 | - | 2 | 482 | 9055728 | ||

Rnase10
Genomic Map