siRNA Details
| Gene Symbol: 4930538K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029198.3 | NP_083474.1 | 7 | 887 | ||
| GenBank mRNA | AK015991.1 | - | 7 | 845 | ||
| AK053970.1 | - | 7 | 888 | |||
| BC048569.1 | - | 7 | 838 | 6705687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930538K18Rik| Gene Symbol: 4930538K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029198.3 | NP_083474.1 | 7 | 887 | ||
| GenBank mRNA | AK015991.1 | - | 7 | 845 | ||
| AK053970.1 | - | 7 | 888 | |||
| BC048569.1 | - | 7 | 838 | 6705687 | ||