siRNA Details
| Gene Symbol: 1700001C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172091.1 | NP_001165562.1 | 2 | 606 | ||
| NM_029296.2 | NP_083572.1 | 4 | 888 | |||
| GenBank mRNA | AK005579.1 | - | 2 | 607 | ||
| BC028263.1 | - | 4 | 851 | 5124927 | ||

1700001C19Rik
Genomic Map