siRNA Details
| Gene Symbol: Morn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029309.2 | NP_083585.1 | 3 | 321 | ||
| GenBank mRNA | AK005808.1 | - | 3 | 309 | ||
| BC036965.1 | - | 3 | 327 | 4511210 | ||
| BC048673.1 | - | 3 | 321 | 6774106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morn5| Gene Symbol: Morn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029309.2 | NP_083585.1 | 3 | 321 | ||
| GenBank mRNA | AK005808.1 | - | 3 | 309 | ||
| BC036965.1 | - | 3 | 327 | 4511210 | ||
| BC048673.1 | - | 3 | 321 | 6774106 | ||