siRNA Details
| Gene Symbol: Fabp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029310.1 | NP_083586.1 | 3 | 337 | ||
| GenBank mRNA | AK005765.1 | - | 2 | 183 | ||
| BC030910.1 | - | 3 | 337 | 4505673 | ||
| EU733649.1 | - | 2 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp12| Gene Symbol: Fabp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029310.1 | NP_083586.1 | 3 | 337 | ||
| GenBank mRNA | AK005765.1 | - | 2 | 183 | ||
| BC030910.1 | - | 3 | 337 | 4505673 | ||
| EU733649.1 | - | 2 | 313 | |||