siRNA Details
| Gene Symbol: Zc3h14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008506.2 | NP_001008506.2 | 15 | 2909 | ||
| NM_001160107.1 | NP_001153579.1 | 14 | 2834 | |||
| NM_001160108.1 | NP_001153580.1 | 7 | 2103 | |||
| NM_029334.2 | NP_083610.2 | 17 | 3302 | |||
| GenBank mRNA | AK048046.1 | - | 17 | 3268 | ||
| AK050226.1 | - | 17 | 3284 | |||
| AK160937.1 | - | 14 | 2820 | |||
| BC024856.1 | - | 10 | 1924 | |||
| BC034760.1 | - | 17 | 3235 | |||

Zc3h14
Genomic Map