siRNA Details
| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029491.2 | NP_083767.2 | 10 | 1481 | ||
| NM_177038.3 | NP_796012.2 | 10 | 1481 | |||
| GenBank mRNA | AK083747.1 | - | 10 | 1481 | ||
| AK173085.1 | - | 3 | 258 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc8| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029491.2 | NP_083767.2 | 10 | 1481 | ||
| NM_177038.3 | NP_796012.2 | 10 | 1481 | |||
| GenBank mRNA | AK083747.1 | - | 10 | 1481 | ||
| AK173085.1 | - | 3 | 258 | |||