siRNA Details
| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029491.2 | NP_083767.2 | 15 | 2093 | ||
| NM_177038.3 | NP_796012.2 | 15 | 2093 | |||
| GenBank mRNA | AK041615.1 | - | 4 | 2180 | ||
| AK083747.1 | - | 15 | 2093 | |||
| AK173085.1 | - | 8 | 870 | |||
| BC048382.1 | - | 4 | 380 | |||

Trappc8
Genomic Map