siRNA Details
| Gene Symbol: CRTAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018058.6 | NP_060528.3 | 15 | 2852 | ||
| GenBank mRNA | AJ276171.1 | - | 15 | 2645 | ||
| AJ279016.1 | - | 14 | 2472 | |||
| AK001182.1 | - | 12 | 2141 | |||
| AK057190.1 | - | 15 | 2852 | |||
| BC034245.1 | - | 13 | 2114 | 4178363 | ||

CRTAC1
Genomic Map