siRNA Details
| Gene Symbol: Clasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081960.1 | NP_001075429.1 | 34 | 4360 | ||
| NM_001114347.1 | NP_001107819.1 | 40 | 5102 | |||
| NM_001286600.1 | NP_001273529.1 | 35 | 4411 | |||
| NM_001286601.1 | NP_001273530.1 | 35 | 4377 | |||
| NM_001286602.1 | NP_001273531.1 | 35 | 4423 | |||
| NM_029633.2 | NP_083909.2 | 33 | 4357 | |||
| GenBank mRNA | AB093252.1 | - | 31 | 3946 | ||
| AJ276961.1 | - | 16 | 2360 | |||
| AK038107.1 | - | 267 | ||||
| AK043551.1 | - | 33 | 4357 | |||
| BC026688.1 | - | 5 | 988 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053722, XM_008766610, XM_008766615, XM_008766611, XM_008766609, and others.

Clasp2
Genomic Map