siRNA Details
| Gene Symbol: Wdr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029687.3 | NP_083963.1 | 7 | 797 | ||
| GenBank mRNA | AK007246.1 | - | 7 | 788 | ||
| AK077027.1 | - | 1045 | ||||
| BC115629.1 | - | 7 | 775 | 40102866 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr38| Gene Symbol: Wdr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029687.3 | NP_083963.1 | 7 | 797 | ||
| GenBank mRNA | AK007246.1 | - | 7 | 788 | ||
| AK077027.1 | - | 1045 | ||||
| BC115629.1 | - | 7 | 775 | 40102866 | ||