siRNA Details
| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029742.2 | NP_084018.1 | 10 | 4342 | ||
| GenBank mRNA | AK046336.1 | - | 1 | 1787 | ||
| AK086657.1 | - | 1 | 181 | |||
| BC056496.1 | - | 1 | 1355 | |||
| BC060132.1 | - | 10 | 4342 | 6409751 | ||

Klhdc10
Genomic Map