siRNA Details
| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029742.2 | NP_084018.1 | 10 | 1675 | ||
| GenBank mRNA | AK039732.1 | - | 9 | 1577 | ||
| AK050289.1 | - | 8 | 1296 | |||
| AK053834.1 | - | 9 | 1547 | |||
| AK085116.1 | - | 9 | 1626 | |||
| AK085130.1 | - | 10 | 1633 | |||
| BC060132.1 | - | 10 | 1675 | 6409751 | ||

Klhdc10
Genomic Map