siRNA Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029752.2 | NP_084028.1 | 3 | 888 | ||
| GenBank mRNA | AK010810.1 | - | 3 | 831 | ||
| AK043288.1 | - | 3 | 830 | |||
| AK044565.1 | - | 3 | 776 | |||
| AK143538.1 | - | 3 | 847 | |||
| BC039941.1 | - | 3 | 846 | 4011957 | ||

Bri3bp
Genomic Map