siRNA Details
| Gene Symbol: Spopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165997.1 | NP_001159469.1 | 12 | 2652 | ||
| NM_001165998.1 | NP_001159470.1 | 11 | 2516 | |||
| NM_029773.2 | NP_084049.2 | 11 | 2555 | |||
| GenBank mRNA | AV228319.2 | - | 1 | 419 | ||

Spopl
Genomic Map