siRNA Details
Gene Symbol: Ccdc116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029779.2 | NP_084055.2 | 4 | 2965 | ||
GenBank mRNA | BC057359.1 | - | 4 | 3312 | 6836034 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029779.2 | NP_084055.2 | 4 | 2965 | ||
GenBank mRNA | BC057359.1 | - | 4 | 3312 | 6836034 |