siRNA Details
| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029779.2 | NP_084055.2 | 4 | 1307 | ||
| GenBank mRNA | AK015290.1 | - | 4 | 1309 | ||
| BC050881.1 | - | 3 | 1321 | 5690792 | ||
| BC057359.1 | - | 4 | 1654 | 6836034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc116| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029779.2 | NP_084055.2 | 4 | 1307 | ||
| GenBank mRNA | AK015290.1 | - | 4 | 1309 | ||
| BC050881.1 | - | 3 | 1321 | 5690792 | ||
| BC057359.1 | - | 4 | 1654 | 6836034 | ||