siRNA Details
| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029779.2 | NP_084055.2 | 4 | 3290 | ||
| GenBank mRNA | BC057359.1 | - | 4 | 3637 | 6836034 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc116| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029779.2 | NP_084055.2 | 4 | 3290 | ||
| GenBank mRNA | BC057359.1 | - | 4 | 3637 | 6836034 | |