siRNA Details
| Gene Symbol: Fbxl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176959.3 | NP_795933.2 | 4 | 1614 | ||
| GenBank mRNA | AK048688.1 | - | 4 | 1629 | ||
| AK129227.1 | - | 4 | 1562 | |||
| AK198158.1 | - | 1 | 16 | |||
| BC091646.1 | - | 4 | 1614 | 30355382 | ||

Fbxl7
Genomic Map