siRNA Details
| Gene Symbol: OR3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002550.2 | NP_002541.2 | 1 | 255 | ||
| XM_017024695.1 | XP_016880184.1 | 2 | 346 | |||
| XM_017024696.1 | XP_016880185.1 | 2 | 320 | |||
| GenBank mRNA | BC069414.1 | - | 1 | 255 | 7262242 | |
| BC096187.3 | - | 1 | 255 | 40004009 | ||
| BC096188.3 | - | 1 | 255 | 40004010 | ||
| BC096189.3 | - | 1 | 255 | 40004011 | ||
| BC096190.1 | - | 1 | 255 | 40004012 | ||

OR3A1
Genomic Map