siRNA Details
| Gene Symbol: Olr1516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000038.1 | NP_001000038.1 | 65 | |||
| Gene Symbol: LOC100909754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750855.1 | XP_003750903.1 | 1 | 65 | ||

Olr1516
Genomic Map