siRNA Details
| Gene Symbol: Olr1561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000048.1 | NP_001000048.1 | 606 | |||
| Gene Symbol: Olr1564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248212.2 | XP_006248274.1 | 1 | 614 | ||

Olr1561
Genomic Map