siRNA Details
| Gene Symbol: RBM28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166135.1 | NP_001159607.1 | 3 | 459 | ||
| NM_018077.2 | NP_060547.2 | 7 | 882 | |||
| XM_011516370.2 | XP_011514672.1 | 8 | 989 | |||
| XM_017012389.1 | XP_016867878.1 | 8 | 990 | |||
| XM_017012390.1 | XP_016867879.1 | 7 | 882 | |||
| GenBank mRNA | AK001239.1 | - | 7 | 882 | ||
| AK222716.1 | - | 7 | 844 | |||
| AK300500.1 | - | 3 | 421 | |||
| BC013889.2 | - | 7 | 836 | 3926178 | ||
| KU178685.1 | - | 7 | 767 | |||

RBM28
Genomic Map