siRNA Details
| Gene Symbol: Olr1704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000271.1 | NP_001000271.1 | 819 | |||
| Gene Symbol: LOC100910894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772701.1 | XP_008770923.1 | 1 | 819 | ||

Olr1704
Genomic Map