siRNA Details
| Gene Symbol: Olr789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000377.1 | NP_001000377.1 | 904 | |||
| Gene Symbol: LOC100909692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749541.1 | XP_003749589.1 | 1 | 904 | ||

Olr789
Genomic Map