siRNA Details
| Gene Symbol: Olr1632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000836.1 | NP_001000836.1 | 526 | |||
| Gene Symbol: LOC103693822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770627.1 | XP_008768849.1 | 1 | 526 | ||
More Information
Set Membership: |
|

Olr1632
Genomic Map