siRNA Details
Gene Symbol: Olr1598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000910.1 | NP_001000910.1 | 1 | 916 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
