siRNA Details
| Gene Symbol: Olr1560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001009.1 | NP_001001009.1 | 1 | 904 | ||
| Gene Symbol: LOC100909884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751033.1 | XP_003751081.1 | 910 | |||

Olr1560
Genomic Map