siRNA Details
| Gene Symbol: Olr1701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001114.1 | NP_001001114.1 | 1 | 937 | ||
| XM_006255861.2 | XP_006255923.1 | 2 | 1128 | |||
| XM_006255863.2 | XP_006255925.1 | 2 | 1097 | |||
| XM_006255865.2 | XP_006255927.1 | 1 | 943 | |||
| Gene Symbol: LOC100910739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751909.2 | XP_003751957.1 | 1 | 748 | ||

Olr1701
Genomic Map