siRNA Details
| Gene Symbol: Olr1701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001114.1 | NP_001001114.1 | 1 | 118 | ||
| XM_006255861.2 | XP_006255923.1 | 2 | 309 | |||
| XM_006255863.2 | XP_006255925.1 | 2 | 278 | |||
| XM_006255865.2 | XP_006255927.1 | 1 | 124 | |||

Olr1701
Genomic Map