siRNA Details
| Gene Symbol: Vom1r89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008928.1 | NP_001008928.1 | 1 | 863 | ||
| GenBank mRNA | AY510298.1 | - | 1 | 863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r89| Gene Symbol: Vom1r89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008928.1 | NP_001008928.1 | 1 | 863 | ||
| GenBank mRNA | AY510298.1 | - | 1 | 863 | ||