siRNA Details
| Gene Symbol: Tuba1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC004745.1 | - | 4 | 1431 | 3583832 | |
| BC022182.1 | - | 4 | 1496 | 3661220 | ||
| BC026753.1 | - | 4 | 1411 | 5124686 | ||
| BC094022.1 | - | 4 | 1886 | 4162287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tuba1c| Gene Symbol: Tuba1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC004745.1 | - | 4 | 1431 | 3583832 | |
| BC022182.1 | - | 4 | 1496 | 3661220 | ||
| BC026753.1 | - | 4 | 1411 | 5124686 | ||
| BC094022.1 | - | 4 | 1886 | 4162287 | ||