siRNA Details
| Gene Symbol: Zfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009570.4 | NP_033596.3 | 8 | 1289 | ||
| GenBank mRNA | AK076618.1 | - | 8 | 1290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfy1| Gene Symbol: Zfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009570.4 | NP_033596.3 | 8 | 1289 | ||
| GenBank mRNA | AK076618.1 | - | 8 | 1290 | ||