siRNA Details
| Gene Symbol: Cyp2c40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010004.2 | NP_034134.2 | 9 | 1597 | ||
| GenBank mRNA | AF047727.1 | - | 9 | 1511 | ||
| BC139471.1 | - | 9 | 1597 | 8862493 | ||
| BC139472.1 | - | 9 | 1597 | 8862494 | ||
| Gene Symbol: Cyp2c68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039555.2 | NP_001034644.2 | 9 | 1565 | ||

Cyp2c40
Genomic Map