siRNA Details
Gene Symbol: CEP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127182.1 | NP_001120654.1 | 2 | 283 | ||
NM_018131.4 | NP_060601.3 | 2 | 405 | |||
XM_011539918.1 | XP_011538220.1 | 2 | 408 | |||
GenBank mRNA | AB091343.1 | - | 2 | 394 | ||
AK223495.1 | - | 2 | 386 | |||
AK315536.1 | - | 2 | 402 | |||
AY788918.1 | - | 2 | 405 | |||
BC008947.2 | - | 2 | 385 | 3028566 | ||
BX640718.1 | - | 3 | 325 | |||
DA545074.1 | - | 2 | 283 |
