siRNA Details
Gene Symbol: Klra16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013794.4 | NP_038822.3 | 7 | 1042 | ||
GenBank mRNA | AF146570.4 | - | 7 | 1042 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klra16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013794.4 | NP_038822.3 | 7 | 1042 | ||
GenBank mRNA | AF146570.4 | - | 7 | 1042 |