siRNA Details
| Gene Symbol: Zmynd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252584.2 | NP_001239513.1 | 11 | 1511 | ||
| NM_001252585.1 | NP_001239514.1 | 10 | 1403 | |||
| NM_001252587.2 | NP_001239516.1 | 12 | 1523 | |||
| NM_001281926.1 | NP_001268855.1 | 11 | 1410 | |||
| NM_001291158.1 | NP_001278087.1 | 12 | 3520 | |||
| NM_027230.5 | NP_081506.3 | 12 | 1598 | |||
| GenBank mRNA | AB093284.2 | - | 12 | 3512 | ||
| AK132427.1 | - | 10 | 1405 | |||
| AK147620.1 | - | 11 | 1411 | |||
| AK155864.1 | - | 11 | 1316 | |||
| BC020059.1 | - | 1 | 323 | |||
| BC023300.1 | - | 11 | 1378 | |||
| BC041797.1 | - | 1 | 326 | |||
| BC048186.1 | - | 12 | 1405 | 6827541 | ||
| BC063267.1 | - | 11 | 1304 | |||

Zmynd8
Genomic Map